2-Amino-3,5-dibromopyrazine 99%

Supplier: APOLLO SCIENTIFIC

Danger

OR01487-25G OR01487-100G
76615-142EA 55 USD
76615-142 76615-144
2-Amino-3,5-dibromopyrazine 99%
2-Amino-3,5-dibromopyrazine
Formula: C₄H₃Br₂N₃
MW: 252.90 g/mol
Melting Pt: 114…117 °C
Storage Temperature: Ambient
MDL Number: MFCD00673150
CAS Number: 24241-18-7
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR