Tantalum(V) ethanolate 99.99%

Supplier: APOLLO SCIENTIFIC

Danger

Synonyms: Tantalum(V) ethoxide

IN3409-10G IN3409-50G
76615-112EA 349 USD
76615-112 76615-110
Tantalum(V) ethanolate 99.99%
Tantalum(V) ethanolate
Formula: C₁₀H₂₅O₅Ta
MW: 406.25 g/mol
Melting Pt: 21 °C
Density: 1.566 g/cm³ (25 °C)
Flash Pt: 30 °C
Storage Temperature: Ambient
MDL Number: MFCD00049785
CAS Number: 6074-84-6
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR