6-Bromo-2,2'-bipyridyl 97%

Supplier: APOLLO SCIENTIFIC

Danger

Synonyms: 6-Bromo-2,2'-bipyridine

OR53209-5G OR53209-1G
76616-958EA 235 USD
76616-958 76616-956
6-Bromo-2,2'-bipyridyl 97%
6-Bromo-2,2'-bipyridyl
Formula: C₁₀H₇BrN₂
MW: 235.0800 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01318953
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR