4-(Methoxycarbonyl)phenylboronic acid 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-(Methoxycarbonyl)benzeneboronic acid(4-(Methoxycarbonyl)phenyl)boronic acid

OR5354-100G OR5354-25G OR5354-5G
76591-700EA 33 USD
76591-700 76591-702 76591-698
4-(Methoxycarbonyl)phenylboronic acid 98%
4-(Methoxycarbonyl)phenylboronic acid
Formula: C₈H₉BO₄
MW: 179.97 g/mol
Melting Pt: 197…200 °C
Storage Temperature: Ambient
MDL Number: MFCD01632203
CAS Number: 99768-12-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR