3-Bromophenyl phenyl ether (BDE No. 2)

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 3-Bromodiphenyl ether1-Bromo-3-phenoxybenzene

OR13134-5G OR13134-1G
76615-616EA 29 USD
76615-616 76615-618
3-Bromophenyl phenyl ether (BDE No. 2)
3-Bromophenyl phenyl ether (BDE No. 2)
Formula: C₁₂H₉BrO
MW: 249.1000 g/mol
Boiling Pt: 156…158 °C/10mmHg
Density: 1.3713 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
UN: 3082
ADR: 9+EHS,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR