4-Methoxyphenylhydrazinium chloride 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Methoxyphenylhydrazine hydrochloride

OR2845-25G OR2845-100G OR2845-5G
76616-236EA 23 USD
76616-236 76616-234 76616-232
4-Methoxyphenylhydrazinium chloride 98%
4-Methoxyphenylhydrazinium chloride
Formula: C₇H₁₁ClN₂O
MW: 174.63 g/mol
Melting Pt: 160…162 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00012945
CAS Number: 19501-58-7
EINECS: 243-115-3
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR