Diisopropyl azodicarboxylate 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Diisopropyl diazene-1,2-dicarboxylate

OR0923-25G OR0923-500G OR0923-100G
76615-278EA 31 USD
76615-278 76615-280 76615-282
Diisopropyl azodicarboxylate 98%
Diisopropyl azodicarboxylate
Formula: C₈H₁₄N₂O₄
MW: 202.21 g/mol
Boiling Pt: 74…75 °C/0.3mmHg
Melting Pt: 44319 °C
Density: 1.027 g/cm³ (25 °C)
Flash Pt: 106 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008875
CAS Number: 2446-83-5
EINECS: 219-502-8
UN: 3082
ADR: 9+EHS,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR