Flurenol (9-hydroxy-9H-fluorene-9-carboxylic acid)

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 9-Hydroxy-9-fluorenecarboxylic acid

OR52146-5G
76625-268EA 21 USD
76625-268
Flurenol (9-hydroxy-9H-fluorene-9-carboxylic acid)
Flurenol (9-hydroxy-9H-fluorene-9-carboxylic acid)
Formula: C₁₄H₁₀O₃
MW: 226.23 g/mol
Melting Pt: 162…166 °C
Storage Temperature: Ambient
MDL Number: MFCD00001137
CAS Number: 467-69-6
UN: 3082
ADR: 9+EHS,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR