2-(Bromomethyl)-1,3-dioxolane 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Bromomethyl-1,3-dioxolane

OR54338-25G OR54338-500G OR54338-100G
76591-788EA 13.39 USD
76591-788 76591-790 76591-792
2-(Bromomethyl)-1,3-dioxolane 97%
2-(Bromomethyl)-1,3-dioxolane
Formula: C₄H₇BrO₂
MW: 167 g/mol
Boiling Pt: 80…82 °C/27mmHg
Density: 1.613 g/cm³ (25 °C)
Flash Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD00003214
CAS Number: 4360-63-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR