Acridine

Supplier: APOLLO SCIENTIFIC
Warning

OR10069-5G OR10069-1G OR10069-10G
76610-892EA 82 USD
76610-892 76610-890 76610-888
Acridine
Acridine
Formula: C₁₃H₉N
Boiling Pt: 346 °C
Melting Pt: 107…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00005025
CAS Number: 260-94-6
EINECS: 205-971-6
UN: 2713
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR