Methyl carbazate

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Methyl hydrazinocarboxylateMethyl hydrazinecarboxylate

OR4850-25G OR4850-100G
76616-866EA 13 USD
76616-866 76616-864
Methyl carbazate
Methyl carbazate
Formula: C₂H₆N₂O₂
MW: 90.08 g/mol
Boiling Pt: 108 °C/12mmHg
Melting Pt: 70…73 °C
Storage Temperature: Ambient
MDL Number: MFCD00007594
CAS Number: 6294-89-9
EINECS: 228-560-3
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR