Dimethyl acetylenedicarboxylate 98%

Supplier: APOLLO SCIENTIFIC

Danger

Synonyms: Dimethyl but-2-yne-1,4-dioateDimethyl but-2-ynedioate

OR1955-250G OR1955-100G OR1955-25G
76620-728EA 19 USD
76620-728 76620-726 76620-724
Dimethyl acetylenedicarboxylate 98%
Dimethyl acetylenedicarboxylate
Formula: C₆H₆O₄
MW: 142.11 g/mol
Boiling Pt: 95…98 °C/19mmHg
Melting Pt: –18 °C
Density: 1.156 g/cm³ (25 °C)
Flash Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00008456
CAS Number: 762-42-5
EINECS: 212-098-4
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR