4-Nitrobenzyl chloroformate 97%

Supplier: APOLLO SCIENTIFIC

Danger

Synonyms: 4-Nitrobenzyl carbonochloridate

OR5386-5G OR5386-100G OR5386-25G
76621-622EA 226 USD
76621-622 76621-624 76621-626
4-Nitrobenzyl chloroformate 97%
4-Nitrobenzyl chloroformate
Formula: C₈H₆ClNO₄
MW: 215.59 g/mol
Boiling Pt: 230 °C/9.75mmHg
Melting Pt: 32…34 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007375
CAS Number: 4457-32-3
EINECS: 224-708-6
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR