Furfurylamine 99%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
Synonyms:
2-(Aminomethyl)furan
OR23026-100G
OR23026-500G
OR23026-25G
76611-332EA
19.57
USD
76611-332
76611-334
76611-336
Furfurylamine 99%
Furfurylamine
Formula:
C₅H₇NO MW: 97.12 g/mol Boiling Pt: 145…146 °C Melting Pt: –70 °C Density: 1.099 g/cm³ (25 °C) Flash Pt: 37 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003258 CAS Number: 617-89-0 EINECS: 210-536-9 UN: 2526 ADR: 3+8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...