Iron(II) trifluoromethanesulfonate

Supplier: APOLLO SCIENTIFIC

Danger

Synonyms: Iron (II) Trifluoromethanesulfonate

IN20952-5G
76615-090EA 68 USD
76615-090
Iron(II) trifluoromethanesulfonate
Iron (II) trifluoromethanesulphonate
Formula: C₂F₆FeO₆S₂
MW: 353.99 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD08460524
UN: 3260
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR