3-Methylcrotonic acid 98%

Supplier: APOLLO SCIENTIFIC

Synonyms: 3,3-Dimethylacrylic acid3-Methylbut-2-enoic acid

OR46069-25G
76621-452EA 13.39 USD
76621-452
3-Methylcrotonic acid 98%
3-Methylcrotonic acid
Formula: C₅H₈O₂
MW: 100.12 g/mol
Boiling Pt: 194…195 °C
Melting Pt: 65…70 °C
Density: 1.969 g/cm³ (25 °C)
Flash Pt: 93.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00004366
CAS Number: 541-47-9
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR