3-Chlorophthalic anhydride, Technical Grade

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Chloroisobenzofuran-1,3-dione

OR54359-5G OR54359-25G OR54359-100G
76591-822EA 16 USD
76591-822 76591-824 76591-820
3-Chlorophthalic anhydride, Technical Grade
3-Chlorophthalic anhydride
Formula: C₈H₃ClO₃
MW: 182.56 g/mol
Melting Pt: 123…126 °C
Storage Temperature: Ambient
MDL Number: MFCD00023107
CAS Number: 117-21-5
EINECS: 204-179-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR