3-Picolyl chloride hydrochloride 97%

Supplier: APOLLO SCIENTIFIC

Danger

Synonyms: 3-(Chloromethyl)pyridine hydrochloride3-(Chloromethyl)-pyridinium chloride

OR0262-100G OR0262-500G OR0262-25G
76615-214EA 42 USD
76615-214 76615-216 76615-218
3-Picolyl chloride hydrochloride 97%
3-Picolyl chloride hydrochloride
Formula: C₆H₇Cl₂N
MW: 164.03 g/mol
Melting Pt: 137…143 °C
Storage Temperature: Ambient
MDL Number: MFCD00012818
CAS Number: 6959-48-4
EINECS: 230-150-4
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR