Piperidine-N-carbaldehyde

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: N-Formylpiperidine1-Formylpiperidine

OR5448-5G
76591-938EA 13.39 USD
76591-938
Piperidine-N-carbaldehyde
Piperidine-N-carbaldehyde
Formula: C₆H₁₁NO
MW: 113.16 g/mol
Boiling Pt: 221…223 °C
Melting Pt: –31 °C
Density: 1.019 g/cm³ (25 °C)
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00006483
CAS Number: 2591-86-8
EINECS: 219-986-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR