4-Aminomorpholine 95%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: N-Aminomorpholine

OR18291-25G OR18291-10G OR18291-5G
76611-256EA 32 USD
76611-256 76611-258 76611-260
4-Aminomorpholine 95%
4-Aminomorpholine
Formula: C₄H₁₀N₂O
Boiling Pt: 167…169 °C
Density: 1.059 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006174
CAS Number: 4319-49-7
EINECS: 224-347-4
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR