Tributylvinyltin

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Tri-n-butylvinyltinTributylvinylstannane

OR42121-5G OR42121-25G OR42121-100G
76611-832EA 303 USD
76611-832 76611-834 76611-836
Tributylvinyltin
Tributylvinyltin
Formula: C₁₄H₃₀Sn
MW: 317.1 g/mol
Boiling Pt: 104…106 °C/3.5mmHg
Density: 1.085 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009421
CAS Number: 7486-35-3
EINECS: 231-291-4
UN: 2788
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR