Methyl-3,4-diaminobenzoate 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Methyl 3,4-diaminobenzoate3,4-Diaminobenzoic acid methyl ester3,4-Diamino benzoate

OR26800-25G OR26800-5G OR26800-100G
76616-186EA 173 USD
76616-186 76616-190 76616-188
Methyl-3,4-diaminobenzoate 98%
Methyl 3,4-diaminobenzoate
Formula: C₈H₁₀N₂O₂
MW: 166.18 g/mol
Melting Pt: 105…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00017098
CAS Number: 36692-49-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR