3-Chloro-2-methylbenzenesulfonyl chloride

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 3-Chloro-2-methylbenzene-1-sulfonyl chloride3-Chloro-2-methylbenzene-1-sulphonyl chloride

OR3972-10G OR3972-25G OR3972-100G
76621-136EA 50 USD
76621-136 76621-132 76621-134
3-Chloro-2-methylbenzenesulfonyl chloride
3-Chloro-2-methylbenzenesulphonyl chloride
Formula: C₇H₆Cl₂O₂S
MW: 225.09 g/mol
Melting Pt: 69…74 °C
Storage Temperature: Ambient
MDL Number: MFCD00052198
CAS Number: 80563-86-6
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR