2-Chloro-5-methyl-3-nitropyridine 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Chloro-3-nitro-5-picoline

OR11054-5G OR11054-100G OR11054-25G
76615-380EA 99 USD
76615-380 76615-382 76615-384
2-Chloro-5-methyl-3-nitropyridine 97%
2-Chloro-5-methyl-3-nitropyridine
Formula: C₆H₅ClN₂O₂
Melting Pt: 46…48 °C
Storage Temperature: Ambient
MDL Number: MFCD02070020
CAS Number: 23056-40-8
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR