4-(Bromomethyl)benzenesulfonyl chloride 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 4-(Bromomethyl)benzene-1-sulfonyl chloride4-(Bromomethyl)benzene-1-sulphonyl chloride

OR5049-5G OR5049-25G
76621-512EA 62 USD
76621-512 76621-514
4-(Bromomethyl)benzenesulfonyl chloride 97%
4-(Bromomethyl)benzenesulphonyl chloride
Formula: C₇H₆BrClO₂S
MW: 269.55 g/mol
Melting Pt: 73…76 °C
Storage Temperature: Ambient
MDL Number: MFCD00156129
CAS Number: 66176-39-4
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR