1-Bromo-3,5-dinitrobenzene 99%

Supplier: APOLLO SCIENTIFIC
Warning

OR307190-5G OR307190-25G OR307190-1G
76625-104EA 32 USD
76625-104 76625-102 76625-100
1-Bromo-3,5-dinitrobenzene 99%
1-Bromo-3,5-dinitrobenzene
Formula: C₆H₃BrN₂O₄
MW: 247 g/mol
Storage Temperature: Ambient
UN: 3459
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR