5-Bromo-2-chlorobenzonitrile

Supplier: APOLLO SCIENTIFIC
Danger

OR4331-25G OR4331-5G OR4331-1G
76625-198EA 16 USD
76625-198 76625-196 76625-194
5-Bromo-2-chlorobenzonitrile
5-Bromo-2-chlorobenzonitrile
Formula: C₇H₃BrClN
MW: 216.46 g/mol
Melting Pt: 133 °C
Storage Temperature: Ambient
MDL Number: MFCD00672948
CAS Number: 57381-44-9
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR