2,4-Dibromothiophene, Technical Grade

Supplier: APOLLO SCIENTIFIC
Warning

OR10849-25G OR10849-5G
76610-970EA 26 USD
76610-970 76610-968
2,4-Dibromothiophene, Technical Grade
2,4-Dibromothiophene
Formula: C₄H₂Br₂S
MW: 241.93 g/mol
Boiling Pt: 215 °C
Density: 2.157 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00043889
CAS Number: 3140-92-9
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR