2-Methoxypyridine 99%

Supplier: APOLLO SCIENTIFIC

Warning

OR13608-100G OR13608-25G OR13608-500G
76611-098EA 15 USD
76611-098 76611-100 76611-096
2-Methoxypyridine 99%
2-Methoxypyridine
Formula: C₆H₇NO
MW: 109.13 g/mol
Boiling Pt: 142…143 °C
Density: 1.049 g/cm³ (25 °C)
Flash Pt: 32 °C
Storage Temperature: Ambient
MDL Number: MFCD00006262
CAS Number: 1628-89-3
EINECS: 216-623-8
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR