3-Bromopyridine 99%

Supplier: APOLLO SCIENTIFIC
Danger

OR23062-500G OR23062-25G OR23062-100G
76616-076EA 108 USD
76616-076 76616-074 76616-072
3-Bromopyridine 99%
3-Bromopyridine
Formula: C₅H₄BrN
MW: 158 g/mol
Boiling Pt: 172…174 °C
Melting Pt: –27 °C
Density: 1.64 g/cm³ (25 °C)
Flash Pt: 51 °C
Storage Temperature: Ambient
MDL Number: MFCD00006373
CAS Number: 626-55-1
EINECS: 210-952-0
UN: 2929
ADR: 6.1+3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR