2-(Bromomethyl)naphthalene, Technical Grade

Supplier: APOLLO SCIENTIFIC

Danger

OR4278-100G OR4278-5G OR4278-25G
76621-366EA 116 USD
76621-366 76621-368 76621-370
2-(Bromomethyl)naphthalene, Technical Grade
2-(Bromomethyl)naphthalene
Formula: C₁₁H₉Br
Boiling Pt: 213 °C/100mmHg
Melting Pt: 52…54 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004123
CAS Number: 939-26-4
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR