(Bromomethyl)cyclopropane 98%

Supplier: APOLLO SCIENTIFIC

Warning

OR28323-5G OR28323-25G OR28323-100G
76611-464EA 68 USD
76611-464 76611-466 76611-468
(Bromomethyl)cyclopropane 98%
(Bromomethyl)cyclopropane
Formula: C₄H₇Br
Boiling Pt: 101…112 °C
Density: 1.392 g/cm³ (25 °C)
Flash Pt: 41 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001306
CAS Number: 7051-34-5
EINECS: 230-331-8
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR