8-Aminoquinoline 98%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: Quinolin-8-ylamine

OR5298-25G OR5298-5G OR5298-100G
76591-470EA 248 USD
76591-470 76591-466 76591-468
8-Aminoquinoline 98%
8-Aminoquinoline
Formula: C₉H₈N₂
MW: 144.18 g/mol
Boiling Pt: 174 °C/26mmHg
Melting Pt: 66…68 °C
Storage Temperature: Ambient
MDL Number: MFCD00006809
CAS Number: 578-66-5
EINECS: 209-427-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR