Mesitylene

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1,3,5-Trimethylbenzene

OR1559-100ML OR1559-2.5L OR1559-500ML
76611-168EA 21 USD
76611-168 76611-170 76611-172
Mesitylene
Mesitylene
Formula: C₉H₁₂
MW: 120.19 g/mol
Boiling Pt: 163…166 °C
Melting Pt: –45 °C
Density: 0.864 g/cm³ (25 °C)
Flash Pt: 53 °C
Storage Temperature: Ambient
MDL Number: MFCD00008538
CAS Number: 108-67-8
EINECS: 203-604-4
UN: 2325
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR