α,α'-Dibromo-p-xylene

Supplier: APOLLO SCIENTIFIC

Danger

Synonyms: p-Xylylene dibromide1,4-Bis(bromomethyl)benzene

OR0107-100G OR0107-25G
76624-628EA 140 USD
76624-628 76624-630
α,α'-Dibromo-p-xylene
α,α'-Dibromo-p-xylene
Formula: C₈H₈Br₂
MW: 263.96 g/mol
Boiling Pt: 156 °C/23mmHg
Melting Pt: 145…147 °C
Density: 1.51 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000182
CAS Number: 623-24-5
EINECS: 210-781-1
UN: 3448
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR