α-Bromo-4-nitrotoluene 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1-(Bromomethyl)-4-nitrobenzeneα-Bromo-p-nitrotoluene4-Nitrobenzyl bromide

OR28642-100G OR28642-500G
76620-942EA 187 USD
76620-942 76620-940
α-Bromo-4-nitrotoluene 98%
α-Bromo-4-nitrotoluene
Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Melting Pt: 97…101 °C
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00007373
CAS Number: 100-11-8
EINECS: 202-820-6
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR