α-Bromo-2,6-dichlorotoluene

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: alpha-Bromo-2,6-dichlorotoluene2,6-Dichlorobenzyl bromide

OR2114-5G
76615-978EA 13.39 USD
76615-978
α-Bromo-2,6-dichlorotoluene
α-Bromo-2,6-dichlorotoluene
Formula: C₇H₅BrCl₂
MW: 239.93 g/mol
Boiling Pt: 135 °C/9.75mmHg
Melting Pt: 54…57 °C
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00000577
CAS Number: 20443-98-5
EINECS: 243-827-4
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR