(S)-(+)-Epichlorhydrin

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: (S)-(+)-3-Chloro-1,2-epoxypropane(2S)-2-(chloromethyl)oxirane(S)-(+)-Glycerol epichlorhydrin(S)-(+)-1-Chloro-2,3-epoxypropane

OR28971-100G OR28971-25G OR28971-5G
76611-530EA 32 USD
76611-530 76611-532 76611-528
(S)-(+)-Epichlorhydrin
(S)-(+)-Epichlorhydrin
Formula: ClCH₂CH(O)CH₂
MW: 92.52 g/mol
Boiling Pt: 114 °C
Density: 1.18 g/cm³ (25 °C)
Flash Pt: 33 °C
Storage Temperature: Ambient
MDL Number: MFCD00077760
CAS Number: 67843-74-7
UN: 2023
ADR: 6.1+3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR