Methyl-3-amino-6-cyclopropylthieno[2,3-b]pyridine-2-carboxylate

Supplier: APOLLO SCIENTIFIC

Warning

OR46756-1G OR46756-250MG
76585-222EA 214 USD
76585-222 76585-220
Methyl-3-amino-6-cyclopropylthieno[2,3-b]pyridine-2-carboxylate
Methyl-3-amino-6-cyclopropylthieno[2,3-b]pyridine-2-carboxylate
Formula: C₁₂H₁₂N₂O₂S
MW: 248.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11841051
CAS Number: 1135283-79-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR