Methyl-2-amino-5-ethyl-1,3-thiazole-4-carboxylate

Supplier: APOLLO SCIENTIFIC
Warning

OR42181-5G OR42181-1G OR42181-250MG
76583-442EA 17 USD
76583-442 76583-444 76583-446
Methyl-2-amino-5-ethyl-1,3-thiazole-4-carboxylate
Methyl-2-amino-5-ethyl-1,3-thiazole-4-carboxylate
Formula: C₇H₁₀N₂O₂S
MW: 186.23 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00728625
CAS Number: 28942-54-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR