1-Boc-isonipecotic acid ethyl ester 95+%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Ethyl N-Boc-piperidine-4-carboxylateEthyl N-BOC-4-piperidinecarboxylate

OR4553-100G OR4553-25G
76584-744EA 57 USD
76584-744 76584-742
1-Boc-isonipecotic acid ethyl ester 95+%
1-Boc-isonipecotic acid ethyl ester
Formula: C₁₃H₂₃NO₄
MW: 257.33 g/mol
Boiling Pt: 120…135 °C/0.5mmHg
Density: 1.046 g/cm³ (25 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD01763998
CAS Number: 142851-03-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR