(5-Bromopyrimidin-2-yl)piperidine 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 5-Bromo-2-(piperidin-1-yl)pyrimidine

OR8548-5G OR8548-1G OR8548-25G
76588-838EA 158 USD
76588-838 76588-836 76588-834
(5-Bromopyrimidin-2-yl)piperidine 98%
(5-Bromopyrimidin-2-yl)piperidine
Formula: C₉H₁₂BrN₃
MW: 242.1157 g/mol
Melting Pt: 48…50 °C
Storage Temperature: Ambient
MDL Number: MFCD00483257
CAS Number: 57356-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR