4-(Benzyloxy)aniline hydrochloride

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Aminophenyl benzyl ether hydrochloride4-Benzyloxyanilinium Chloridep-Benzyloxyaniline hydrochloride

OR8024-25G
76588-510EA 13.39 USD
76588-510
4-(Benzyloxy)aniline hydrochloride
4-(Benzyloxy)aniline hydrochloride
Formula: C₁₃H₁₄ClNO
Melting Pt: 227…229 °C
Storage Temperature: Ambient
MDL Number: MFCD00012995
CAS Number: 51388-20-6
EINECS: 257-170-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR