Methyl-5-hydroxypicolinate 95+%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Methyl 5-hydroxypyridine-2-carboxylate

OR51766-1G OR51766-5G
76586-712EA 157.59 USD
76586-712 76586-710
Methyl-5-hydroxypicolinate 95+%
Methyl 5-hydroxypicolinate
Formula: C₇H₇NO₃
MW: 153.1354 g/mol
Melting Pt: 191…197 °C
Storage Temperature: Ambient
MDL Number: MFCD05256426
CAS Number: 30766-12-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR