4-(N,N-Dimethylamino)phenylboronic acid 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-(Dimethylamino)benzeneboronic acid

OR7205-25G OR7205-5G OR7205-1G
76587-770EA 63 USD
76587-770 76587-772 76587-774
4-(N,N-Dimethylamino)phenylboronic acid 98%
4-(N,N-Dimethylamino)phenylboronic acid
Formula: C₈H₁₂BNO₂
MW: 165.00
Melting Pt: 227 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01074642
CAS Number: 28611-39-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR