4-Isoquinolinol

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 4-HydroxyisoquinolineIsoquinolin-4-ol

OR48080-5G OR48080-250MG OR48080-1G
76585-658EA 31 USD
76585-658 76585-662 76585-660
4-Isoquinolinol
4-Isoquinolinol
Formula: C₉H₇NO
MW: 145.1580 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00234514
CAS Number: 3336-49-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR