Methyl-3,5-dibromo-4-methylbenzoate

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: Methyl 3,5-dibromo-4-methylbenzoate

OR4685-100G OR4685-25G
76585-284EA 44 USD
76585-284 76585-286
Methyl-3,5-dibromo-4-methylbenzoate
Methyl 3,5-dibromo-4-methylbenzoate
Formula: C₉H₈Br₂O₂
MW: 307.98 g/mol
Melting Pt: 86…90 °C
Storage Temperature: Ambient
MDL Number: MFCD00010729
CAS Number: 74896-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR