3-Methyl-4-isoxazolecarboxylic acid

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 3-Methylisoxazole-4-carboxylic acid

OR7097-250MG OR7097-1G OR7097-5G
76587-660EA 41 USD
76587-660 76587-662 76587-664
3-Methyl-4-isoxazolecarboxylic acid
3-Methyl-4-isoxazolecarboxylic acid
Formula: C₅H₅NO₃
MW: 127.10 g/mol
Melting Pt: 183…190 °C
Storage Temperature: Ambient
MDL Number: MFCD01318161
CAS Number: 17153-20-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR