3-Amino-2-bromo-4,6-dimethylpyridine 97%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 2-Bromo-4,6-dimethylpyridin-3-amine

OR7028-1G OR7028-500MG OR7028-250MG
76587-584EA 58 USD
76587-584 76587-580 76587-582
3-Amino-2-bromo-4,6-dimethylpyridine 97%
3-Amino-2-bromo-4,6-dimethylpyridine
Formula: C₇H₉BrN₂
MW: 201.0600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03731181
CAS Number: 104829-98-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR