Methyl-2-chloro-6-methylisonicotinate 97%

Supplier: APOLLO SCIENTIFIC

Danger

Synonyms: Methyl 2-chloro-6-picoline-4-carboxylateMethyl 2-chloro-6-methylpyridine-4-carboxylate

OR8630-5G OR8630-1G
76588-944EA 27 USD
76588-944 76588-942
Methyl-2-chloro-6-methylisonicotinate 97%
Methyl 2-chloro-6-methylisonicotinate
Formula: C₈H₈ClNO₂
MW: 185.61 g/mol
Boiling Pt: 124…125 °C/7mmHg
Melting Pt: 58…62 °C
Storage Temperature: Ambient
MDL Number: MFCD04038391
CAS Number: 3998-90-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR